gRNA Design Details
- Target DNA Sequence
- CCTCTTGCTAATTCAGTTTT
- PAM Sequence
- TGG
- Target locus
-
Chr.6: 49404823 - 49404845 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
79.92/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Stk31
- Entrez Gene ID
-
77485
- Gene Name
- serine threonine kinase 31
- Gene Aliases
-
C330007K24Rik
- Chromosome Location
-
Chr.6: 49395483 - 49469506 on GRCm38
- UniGene ID
- Mm.128054
- Species
-
Mouse