gRNA Design Details
- Target DNA Sequence
- GTTGGCTACGCAGGTAGTCC
- PAM Sequence
- AGG
- Target locus
-
Chr.7: 139122842 - 139122864 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
86.71/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Stk32c
- Entrez Gene ID
-
57740
- Gene Name
- serine/threonine kinase 32C
- Gene Aliases
-
PKE, Pkek, YANK3
- Chromosome Location
-
Chr.7: 139103638 - 139213307 on GRCm38
- UniGene ID
- Mm.65336
- Species
-
Mouse