gRNA Design Details
- Target DNA Sequence
- CTCCCCATCCTTTATGATGG
- PAM Sequence
- TGG
- Target locus
-
Chr.7: 139121798 - 139121820 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
84.84/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Stk32c
- Entrez Gene ID
-
57740
- Gene Name
- serine/threonine kinase 32C
- Gene Aliases
-
PKE, Pkek, YANK3
- Chromosome Location
-
Chr.7: 139103638 - 139213307 on GRCm38
- UniGene ID
- Mm.65336
- Species
-
Mouse