gRNA Design Details
- Target DNA Sequence
- GGTACACTGACGTGTCCCAA
- PAM Sequence
- AGG
- Target locus
-
Chr.18: 43967035 - 43967057 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
91.82/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Spink5
- Entrez Gene ID
-
72432
- Gene Name
- serine peptidase inhibitor, Kazal type 5
- Gene Aliases
-
2310065D10Rik, AV238912, LEKT1, LETK1, VAKT1
- Chromosome Location
-
Chr.18: 43959545 - 44022487 on GRCm38
- UniGene ID
- Mm.35369
- Species
-
Mouse