gRNA Design Details
- Target DNA Sequence
- GGTCAGACCAGTTTACAACT
- PAM Sequence
- GGG
- Target locus
-
Chr.9: 48959254 - 48959276 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
87.27/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Htr3b
- Entrez Gene ID
-
57014
- Gene Name
- 5-hydroxytryptamine (serotonin) receptor 3B
- Gene Aliases
-
5-HT3B
- Chromosome Location
-
Chr.9: 48935008 - 48964990 on GRCm38
- UniGene ID
- Mm.117131
- Species
-
Mouse