gRNA Design Details
- Target DNA Sequence
- GTCTATCCAGCCGGAGGAGA
- PAM Sequence
- AGG
- Target locus
-
Chr.9: 4309301 - 4309323 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
85.42/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Aasdhppt
- Entrez Gene ID
-
67618
- Gene Name
- aminoadipate-semialdehyde dehydrogenase-phosphopantetheinyl transferase
- Gene Aliases
-
2010309J24Rik, 2810407B07Rik, AASD-PPT, CGI-80, LYS2, LYS5
- Chromosome Location
-
Chr.9: 4294793 - 4309509 on GRCm38
- UniGene ID
- Mm.33970
- Species
-
Mouse