gRNA Design Details
- Target DNA Sequence
- GGAAACGGGACTCGGTAAAA
- PAM Sequence
- TGG
- Target locus
-
Chr.10: 117078842 - 117078864 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
87.96/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Frs2
- Entrez Gene ID
-
327826
- Gene Name
- fibroblast growth factor receptor substrate 2
- Gene Aliases
-
4732458E18, C330018A15Rik, Frs2alpha, SNT1
- Chromosome Location
-
Chr.10: 117070127 - 117148474 on GRCm38
- UniGene ID
- Mm.135965
- Species
-
Mouse