gRNA Design Details

Target DNA Sequence
GGAAACGGGACTCGGTAAAA
PAM Sequence
TGG
Target locus
Chr.10: 117078842 - 117078864 on GRCm38
Strand
Reverse
Application
Gene Knock-Out
Score: 87.96/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Frs2
Entrez Gene ID
327826
Gene Name
fibroblast growth factor receptor substrate 2
Gene Aliases
4732458E18, C330018A15Rik, Frs2alpha, SNT1
Chromosome Location
Chr.10: 117070127 - 117148474 on GRCm38
UniGene ID
Mm.135965
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_177798.3 NP_808466.1 7