gRNA Design Details
- Target DNA Sequence
- ACTCCAGATCATGCAGGGCC
- PAM Sequence
- TGG
- Target locus
-
Chr.18: 43243080 - 43243102 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
84.6/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Stk32a
- Entrez Gene ID
-
269019
- Gene Name
- serine/threonine kinase 32A
- Gene Aliases
-
A930015B13Rik, YANK1
- Chromosome Location
-
Chr.18: 43207697 - 43317481 on GRCm38
- UniGene ID
- Mm.150401
- Species
-
Mouse