gRNA Design Details
- Target DNA Sequence
- GTATTCCTTCCAAGATGAGG
- PAM Sequence
- AGG
- Target locus
-
Chr.18: 43261248 - 43261270 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
83.18/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Stk32a
- Entrez Gene ID
-
269019
- Gene Name
- serine/threonine kinase 32A
- Gene Aliases
-
A930015B13Rik, YANK1
- Chromosome Location
-
Chr.18: 43207697 - 43317481 on GRCm38
- UniGene ID
- Mm.150401
- Species
-
Mouse