gRNA Design Details

Target DNA Sequence
GTATTCCTTCCAAGATGAGG
PAM Sequence
AGG
Target locus
Chr.18: 43261248 - 43261270 on GRCm38
Strand
Forward
Application
Gene Knock-Out
Score: 83.18/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Stk32a
Entrez Gene ID
269019
Gene Name
serine/threonine kinase 32A
Gene Aliases
A930015B13Rik, YANK1
Chromosome Location
Chr.18: 43207697 - 43317481 on GRCm38
UniGene ID
Mm.150401
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_178749.3 NP_848864.1 5