gRNA Design Details
- Target DNA Sequence
- TTTTCGAGGACGCTCGCGTT
- PAM Sequence
- CGG
- Target locus
-
Chr.19: 6950728 - 6950750 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
91/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Bad
- Entrez Gene ID
-
12015
- Gene Name
- BCL2-associated agonist of cell death
- Gene Aliases
-
AI325008, Bbc2
- Chromosome Location
-
Chr.19: 6941861 - 6951905 on GRCm38
- UniGene ID
- Mm.4387
- Species
-
Mouse