gRNA Design Details
- Target DNA Sequence
- CCGGAAATGTACTGGTGATT
- PAM Sequence
- TGG
- Target locus
-
Chr.1: 134234260 - 134234282 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
85.75/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Adora1
- Entrez Gene ID
-
11539
- Gene Name
- adenosine A1 receptor
- Gene Aliases
-
A1-AR, A1AR, A1R, AA1R, AI848715, BB176431, Ri
- Chromosome Location
-
Chr.1: 134199215 - 134235457 on GRCm38
- UniGene ID
- Mm.298908
- Species
-
Mouse