gRNA Design Details
- Target DNA Sequence
- ATGTCGTCCGGGTCCAAGGA
- PAM Sequence
- AGG
- Target locus
-
Chr.7: 4691080 - 4691102 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
87.19/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Brsk1
- Entrez Gene ID
-
381979
- Gene Name
- BR serine/threonine kinase 1
- Gene Aliases
-
Gm1100, SAD-B, SADB
- Chromosome Location
-
Chr.7: 4690928 - 4715997 on GRCm38
- UniGene ID
- Mm.297064
- Species
-
Mouse