gRNA Design Details
- Target DNA Sequence
- TGGCTTGGTGACGGGTATGA
- PAM Sequence
- GGG
- Target locus
-
Chr.10: 75333033 - 75333055 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
91.82/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Adora2a
- Entrez Gene ID
-
11540
- Gene Name
- adenosine A2a receptor
- Gene Aliases
-
A2AAR, A2aR, AA2AR
- Chromosome Location
-
Chr.10: 75316860 - 75334792 on GRCm38
- UniGene ID
- Mm.333734
- Species
-
Mouse