gRNA Design Details
- Target DNA Sequence
- ATACTTCAGCAGCGTGGAGG
- PAM Sequence
- AGG
- Target locus
-
Chr.5: 30113333 - 30113355 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
85.68/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Gareml
- Entrez Gene ID
-
242915
- Gene Name
- GRB2 associated, regulator of MAPK1-like
- Gene Aliases
-
Fam59b, Gm444
- Chromosome Location
-
Chr.5: 30105196 - 30118380 on GRCm38
- UniGene ID
- Mm.332287
- Species
-
Mouse