gRNA Design Details
- Target DNA Sequence
- AGGGTTTAGTGAAGCCCCGA
- PAM Sequence
- GGG
- Target locus
-
Chr.9: 110798311 - 110798333 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
86.64/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Prss42
- Entrez Gene ID
-
235628
- Gene Name
- protease, serine 42
- Gene Aliases
-
Tessp2
- Chromosome Location
-
Chr.9: 110797261 - 110803744 on GRCm38
- UniGene ID
- Mm.292285
- Species
-
Mouse