gRNA Design Details
- Target DNA Sequence
- GTTCACTGCCCTTCTTATGG
- PAM Sequence
- TGG
- Target locus
-
Chr.6: 61311064 - 61311086 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
85.35/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Ccser1
- Entrez Gene ID
-
232035
- Gene Name
- coiled-coil serine rich 1
- Gene Aliases
-
6230405M12Rik, C130092O11Rik, Fam190a
- Chromosome Location
-
Chr.6: 61180325 - 62382863 on GRCm38
- UniGene ID
- Mm.150324
- Species
-
Mouse