gRNA Design Details
- Target DNA Sequence
- TTCGGTTTTGGTCACGTTGT
- PAM Sequence
- TGG
- Target locus
-
Chr.2: 58315842 - 58315864 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
92.36/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Acvr1c
- Entrez Gene ID
-
269275
- Gene Name
- activin A receptor, type IC
- Gene Aliases
-
ACVRLK7, ALK7, Alk-7, C230097P10
- Chromosome Location
-
Chr.2: 58267453 - 58357896 on GRCm38
- UniGene ID
- Mm.77751
- Species
-
Mouse