gRNA Design Details
- Target DNA Sequence
- CTTCGGTCAACAGATGCCGG
- PAM Sequence
- TGG
- Target locus
-
Chr.15: 74672216 - 74672238 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
96.73/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Arc
- Entrez Gene ID
-
11838
- Gene Name
- activity regulated cytoskeletal-associated protein
- Gene Aliases
-
Arc3.1, C86064, arg3.1, mArc
- Chromosome Location
-
Chr.15: 74669081 - 74672570 on GRCm38
- UniGene ID
- Mm.491310
- Species
-
Mouse