gRNA Design Details
- Target DNA Sequence
- TCACTCAGAGACAGCCAGAT
- PAM Sequence
- GGG
- Target locus
-
Chr.6: 13215754 - 13215776 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
83.68/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Vwde
- Entrez Gene ID
-
232585
- Gene Name
- von Willebrand factor D and EGF domains
- Gene Aliases
-
Gm467
- Chromosome Location
-
Chr.6: 13185610 - 13224965 on GRCm38
- UniGene ID
- Mm.389082
- Species
-
Mouse