gRNA Design Details
- Target DNA Sequence
- AGATCAGAGGGCCGATTTCC
- PAM Sequence
- AGG
- Target locus
-
Chr.18: 44080127 - 44080149 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
85.82/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Spink6
- Entrez Gene ID
-
433180
- Gene Name
- serine peptidase inhibitor, Kazal type 6
- Gene Aliases
-
EG433180
- Chromosome Location
-
Chr.18: 44071393 - 44083610 on GRCm38
- UniGene ID
- Mm.57302
- Species
-
Mouse