gRNA Design Details
- Target DNA Sequence
- GGCTCTGATGGCCAGACATA
- PAM Sequence
- TGG
- Target locus
-
Chr.5: 148213973 - 148213995 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
68.28/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- SPINK6
- Entrez Gene ID
-
404203
- Gene Name
- serine peptidase inhibitor, Kazal type 6
- Gene Aliases
-
BUSI2, UNQ844
- Chromosome Location
-
Chr.5: 148202794 - 148215137 on GRCh38
- UniGene ID
- Hs.334274
- Species
-
Human