gRNA Design Details
- Target DNA Sequence
- TGTACTGTGGGGATTTCCGG
- PAM Sequence
- AGG
- Target locus
-
Chr.2: 26589340 - 26589362 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
88.42/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Egfl7
- Entrez Gene ID
-
353156
- Gene Name
- EGF-like domain 7
- Gene Aliases
-
VE-statin, Zneu1
- Chromosome Location
-
Chr.2: 26581028 - 26592682 on GRCm38
- UniGene ID
- Mm.268933
- Species
-
Mouse