gRNA Design Details
- Target DNA Sequence
- ATCCGTCGTAGCAGGTACAC
- PAM Sequence
- CGG
- Target locus
-
Chr.17: 28153629 - 28153651 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
92.64/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Scube3
- Entrez Gene ID
-
268935
- Gene Name
- signal peptide, CUB domain, EGF-like 3
- Gene Aliases
-
CEGF3, D030038I21Rik
- Chromosome Location
-
Chr.17: 28142100 - 28174851 on GRCm38
- UniGene ID
- Mm.258491
- Species
-
Mouse