gRNA Design Details

Target DNA Sequence
ATCCGTCGTAGCAGGTACAC
PAM Sequence
CGG
Target locus
Chr.17: 28153629 - 28153651 on GRCm38
Strand
Reverse
Application
Gene Knock-Out
Score: 92.64/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Scube3
Entrez Gene ID
268935
Gene Name
signal peptide, CUB domain, EGF-like 3
Gene Aliases
CEGF3, D030038I21Rik
Chromosome Location
Chr.17: 28142100 - 28174851 on GRCm38
UniGene ID
Mm.258491
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001004366.1 NP_001004366.1 3