gRNA Design Details

Target DNA Sequence
GTCTCAGCGAAGCAGCTCGA
PAM Sequence
TGG
Target locus
Chr.2: 31778376 - 31778398 on GRCm38
Strand
Forward
Application
Gene Knock-Out
Score: 87.94/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Abl1
Entrez Gene ID
11350
Gene Name
c-abl oncogene 1, non-receptor tyrosine kinase
Gene Aliases
AI325092, Abl, E430008G22Rik, c-Abl
Chromosome Location
Chr.2: 31688252 - 31807093 on GRCm38
UniGene ID
Mm.1318
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001112703.2 NP_001106174.1 2
NM_001283045.1 NP_001269974.1 2
NM_001283046.1 NP_001269975.1 3
NM_001283047.1 NP_001269976.1 3
NM_009594.4 NP_033724.2 2