gRNA Design Details
- Target DNA Sequence
- TATATCGACAGCTGCCACGG
- PAM Sequence
- AGG
- Target locus
-
Chr.14: 33944670 - 33944692 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
92.11/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Gdf2
- Entrez Gene ID
-
12165
- Gene Name
- growth differentiation factor 2
- Gene Aliases
-
Bmp9
- Chromosome Location
-
Chr.14: 33941039 - 33947198 on GRCm38
- UniGene ID
- Mm.422844
- Species
-
Mouse