gRNA Design Details
- Target DNA Sequence
- AAGTCGATCATGTACTGGGG
- PAM Sequence
- TGG
- Target locus
-
Chr.14: 33941400 - 33941422 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
91.43/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Gdf2
- Entrez Gene ID
-
12165
- Gene Name
- growth differentiation factor 2
- Gene Aliases
-
Bmp9
- Chromosome Location
-
Chr.14: 33941039 - 33947198 on GRCm38
- UniGene ID
- Mm.422844
- Species
-
Mouse