gRNA Design Details
- Target DNA Sequence
- TGAAGGCTTGCACACAAGGC
- PAM Sequence
- TGG
- Target locus
-
Chr.4: 154177084 - 154177106 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
82.58/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Megf6
- Entrez Gene ID
-
230971
- Gene Name
- multiple EGF-like-domains 6
- Gene Aliases
-
2600001P17Rik, Egfl3
- Chromosome Location
-
Chr.4: 154170713 - 154275721 on GRCm38
- UniGene ID
- Mm.275216
- Species
-
Mouse