gRNA Design Details

Target DNA Sequence
GCCACTTGTGCTAAGCTGTA
PAM Sequence
AGG
Target locus
Chr.17: 34598184 - 34598206 on GRCm38
Strand
Forward
Application
Gene Knockout
Score: 90.91/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Ager
Entrez Gene ID
11596
Gene Name
advanced glycosylation end product-specific receptor
Gene Aliases
RAGE
Chromosome Location
Chr.17: 34597410 - 34600937 on GRCm38
UniGene ID
Mm.3383
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001271422.1 NP_001258351.1 2
NM_001271423.1 NP_001258352.1 2
NM_001271424.1 NP_001258353.1 1
NM_007425.3 NP_031451.2 2
NR_073173.1 2
NR_073175.1 1
NR_073176.1 1