gRNA Design Details
- Target DNA Sequence
- GGTCAAGTCACAGTACACCG
- PAM Sequence
- AGG
- Target locus
-
Chr.11: 28854515 - 28854537 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
93.73/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Efemp1
- Entrez Gene ID
-
216616
- Gene Name
- epidermal growth factor-containing fibulin-like extracellular matrix protein 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.11: 28853205 - 28926743 on GRCm38
- UniGene ID
- Mm.44176
- Species
-
Mouse