gRNA Design Details
- Target DNA Sequence
- CATCTTTAACCTGGATGGCG
- PAM Sequence
- TGG
- Target locus
-
Chr.15: 101194005 - 101194027 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
87.28/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Acvr1b
- Entrez Gene ID
-
11479
- Gene Name
- activin A receptor, type 1B
- Gene Aliases
-
6820432J04, ActR-IB, ActRIB, Acvrlk4, Alk4, SKR2
- Chromosome Location
-
Chr.15: 101174125 - 101212601 on GRCm38
- UniGene ID
- Mm.308467
- Species
-
Mouse