gRNA Design Details
- Target DNA Sequence
- ACCGACCTCAAGCTCTTGCG
- PAM Sequence
- TGG
- Target locus
-
Chr.1: 133066525 - 133066547 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
91.55/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Pik3c2b
- Entrez Gene ID
-
240752
- Gene Name
- phosphoinositide-3-kinase, class 2, beta polypeptide
- Gene Aliases
-
C330011J12Rik, Gm199, PI3K-C2beta, PI3KC2beta
- Chromosome Location
-
Chr.1: 133045672 - 133108688 on GRCm38
- UniGene ID
- Mm.387308
- Species
-
Mouse