gRNA Design Details
- Target DNA Sequence
- TCTCGTATAGCTTGCGTTTT
- PAM Sequence
- CGG
- Target locus
-
Chr.12: 113544214 - 113544236 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
84.05/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Adam6a
- Entrez Gene ID
-
238406
- Gene Name
- a disintegrin and metallopeptidase domain 6A
- Gene Aliases
-
Adam6, tMDCIV
- Chromosome Location
-
Chr.12: 113543908 - 113546414 on GRCm38
- UniGene ID
- Mm.188683
- Species
-
Mouse