gRNA Design Details
- Target DNA Sequence
- CGGGACTCTATAGATGCCAT
- PAM Sequence
- AGG
- Target locus
-
Chr.4: 148082275 - 148082297 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
86.77/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Agtrap
- Entrez Gene ID
-
11610
- Gene Name
- angiotensin II, type I receptor-associated protein
- Gene Aliases
-
3300002E14Rik, AT1R, Atrap, D4Wsu124e
- Chromosome Location
-
Chr.4: 148077061 - 148088064 on GRCm38
- UniGene ID
- Mm.46247
- Species
-
Mouse