gRNA Design Details
- Target DNA Sequence
- TGCTGATAGATGCCCAGCCA
- PAM Sequence
- GGG
- Target locus
-
Chr.11: 77498966 - 77498988 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
83.42/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Git1
- Entrez Gene ID
-
216963
- Gene Name
- G protein-coupled receptor kinase-interactor 1
- Gene Aliases
-
Cat-1, p95Cat
- Chromosome Location
-
Chr.11: 77493412 - 77507775 on GRCm38
- UniGene ID
- Mm.290182
- Species
-
Mouse