gRNA Design Details
- Target DNA Sequence
- ATCGGAAGGGTCTTCGGCAT
- PAM Sequence
- CGG
- Target locus
-
Chr.6: 86850155 - 86850177 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
91.82/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Aak1
- Entrez Gene ID
-
269774
- Gene Name
- AP2 associated kinase 1
- Gene Aliases
-
5530400K14Rik, 9630042K20, AU067724, AU067726, BC028270, C79663, D6Ertd245e, R75501, mKIAA1048
- Chromosome Location
-
Chr.6: 86849482 - 87003227 on GRCm38
- UniGene ID
- Mm.221038
- Species
-
Mouse