gRNA Design Details

Target DNA Sequence
GTTGACAAACATACGTTTCA
PAM Sequence
AGG
Target locus
Chr.6: 86925065 - 86925087 on GRCm38
Strand
Reverse
Application
Gene Knockout
Score: 82.73/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Aak1
Entrez Gene ID
269774
Gene Name
AP2 associated kinase 1
Gene Aliases
5530400K14Rik, 9630042K20, AU067724, AU067726, BC028270, C79663, D6Ertd245e, R75501, mKIAA1048
Chromosome Location
Chr.6: 86849482 - 87003227 on GRCm38
UniGene ID
Mm.221038
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001040106.2 NP_001035195.1 2
NM_177762.6 NP_808430.2 2