gRNA Design Details
- Target DNA Sequence
- CAGGTCATCCCACGAGCTCT
- PAM Sequence
- TGG
- Target locus
-
Chr.17: 23834792 - 23834814 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
82.18/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Prss33
- Entrez Gene ID
-
353130
- Gene Name
- protease, serine 33
- Gene Aliases
-
Eos, mT6, tryptase-6
- Chromosome Location
-
Chr.17: 23833338 - 23836126 on GRCm38
- UniGene ID
- Mm.261689
- Species
-
Mouse