gRNA Design Details
- Target DNA Sequence
- GATCGGACTTCCTCAAGCCT
- PAM Sequence
- AGG
- Target locus
-
Chr.19: 6942364 - 6942386 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
87.91/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Bad
- Entrez Gene ID
-
12015
- Gene Name
- BCL2-associated agonist of cell death
- Gene Aliases
-
AI325008, Bbc2
- Chromosome Location
-
Chr.19: 6941861 - 6951905 on GRCm38
- UniGene ID
- Mm.4387
- Species
-
Mouse