gRNA Design Details
- Target DNA Sequence
- ATAGAATCCCACGTCCCATT
- PAM Sequence
- GGG
- Target locus
-
Chr.1: 65336997 - 65337019 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
87.72/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Pth2r
- Entrez Gene ID
-
213527
- Gene Name
- parathyroid hormone 2 receptor
- Gene Aliases
-
Pthr2
- Chromosome Location
-
Chr.1: 65280282 - 65389625 on GRCm38
- UniGene ID
- Mm.294225
- Species
-
Mouse