gRNA Design Details
- Target DNA Sequence
- CTGCAGCCTGATATCAACAT
- PAM Sequence
- AGG
- Target locus
-
Chr.1: 65337063 - 65337085 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
86.34/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Pth2r
- Entrez Gene ID
-
213527
- Gene Name
- parathyroid hormone 2 receptor
- Gene Aliases
-
Pthr2
- Chromosome Location
-
Chr.1: 65280282 - 65389625 on GRCm38
- UniGene ID
- Mm.294225
- Species
-
Mouse