gRNA Design Details
- Target DNA Sequence
- GATGTCGCTTTACAGGAGCC
- PAM Sequence
- TGG
- Target locus
-
Chr.12: 8301648 - 8301670 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
88.6/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Gdf7
- Entrez Gene ID
-
238057
- Gene Name
- growth differentiation factor 7
- Gene Aliases
-
BMP12
- Chromosome Location
-
Chr.12: 8291018 - 8302088 on GRCm38
- UniGene ID
- Mm.271308
- Species
-
Mouse