gRNA Design Details
- Target DNA Sequence
- ACACCAAGTGCCGCGTACAA
- PAM Sequence
- AGG
- Target locus
-
Chr.2: 31760411 - 31760433 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
91.82/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Abl1
- Entrez Gene ID
-
11350
- Gene Name
- c-abl oncogene 1, non-receptor tyrosine kinase
- Gene Aliases
-
AI325092, Abl, E430008G22Rik, c-Abl
- Chromosome Location
-
Chr.2: 31688252 - 31807093 on GRCm38
- UniGene ID
- Mm.1318
- Species
-
Mouse