gRNA Design Details
- Target DNA Sequence
- TAAAGTGGTGCTTCCTGGTA
- PAM Sequence
- GGG
- Target locus
-
Chr.2: 139537601 - 139537623 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
83.42/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Sptlc3
- Entrez Gene ID
-
228677
- Gene Name
- serine palmitoyltransferase, long chain base subunit 3
- Gene Aliases
-
C130053K05Rik
- Chromosome Location
-
Chr.2: 139493913 - 139637674 on GRCm38
- UniGene ID
- Mm.100450
- Species
-
Mouse