gRNA Design Details
- Target DNA Sequence
- GATCCATATCAACAAGATCT
- PAM Sequence
- TGG
- Target locus
-
Chr.1: 59813623 - 59813645 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
83.53/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Bmpr2
- Entrez Gene ID
-
12168
- Gene Name
- bone morphogenetic protein receptor, type II (serine/threonine kinase)
- Gene Aliases
-
2610024H22Rik, AL117858, AW546137, BB189135, BMP-2, BMPR-2, BMPR-II, BMPRII, BRK-3, Gm20272
- Chromosome Location
-
Chr.1: 59764270 - 59878081 on GRCm38
- UniGene ID
- Mm.391654
- Species
-
Mouse