gRNA Design Details

Target DNA Sequence
GACAGCCAAATGATCCTCGT
PAM Sequence
TGG
Target locus
Chr.2: 175121511 - 175121533 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 68.04/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
ATF2
Entrez Gene ID
1386
Gene Name
activating transcription factor 2
Gene Aliases
CRE-BP1, CREB-2, CREB2, HB16, TREB7
Chromosome Location
Chr.2: 175072250 - 175168206 on GRCh38
UniGene ID
Hs.592510
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001256090.1 NP_001243019.1 6
NM_001256091.1 NP_001243020.1 5
NM_001256094.1 NP_001243023.1 5
NM_001880.3 NP_001871.2 5
NR_045768.1 6
NR_045769.1 6
NR_045770.1 6
NR_045771.1 5
NR_045774.1 5