gRNA Design Details
- Target DNA Sequence
- CTGTTTCCGACCCATCAGAG
- PAM Sequence
- AGG
- Target locus
-
Chr.9: 86755226 - 86755248 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
86.14/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Prss35
- Entrez Gene ID
-
244954
- Gene Name
- protease, serine 35
- Gene Aliases
-
6030424L22Rik, P3D9
- Chromosome Location
-
Chr.9: 86743240 - 86758443 on GRCm38
- UniGene ID
- Mm.257629
- Species
-
Mouse