gRNA Design Details
- Target DNA Sequence
- GTGCTTGGCACTGGAATCGC
- PAM Sequence
- AGG
- Target locus
-
Chr.13: 110354020 - 110354042 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
91.57/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Gapt
- Entrez Gene ID
-
238875
- Gene Name
- Grb2-binding adaptor, transmembrane
- Gene Aliases
-
9830130M13Rik
- Chromosome Location
-
Chr.13: 110352614 - 110357273 on GRCm38
- UniGene ID
- Mm.30533
- Species
-
Mouse